missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Hooded Protective Coveralls 4510 Series, Elastic Wrist and Ankles
Coveralls provide light duty protection against infective agents, certain hazardous dusts, and limited liquid splashes. PE laminate material includes micropores that allow heat and moisture to evaporate from the skin and out through the suit for a lightweight and comfortable fit.
159.00 € - 164.00 €
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Material | Microporous Polyethylene Laminate |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11963734
|
3M™
7000089662 |
Medium |
164.00 €
Pack of 20 |
Estimated Shipment: 26-03-2025
Log in to see stock. |
|||||
11983734
|
3M™
7000089663 |
Large |
159.00 €
Pack of 20 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
12972315
|
3M™
7000089664 |
X-Large |
164.00 €
Pack of 20 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
12982315
|
3M™
7000089665 |
2X-Large |
159.00 €
Pack of 20 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
12992315
|
3M™
7000089666 |
3X-Large |
159.00 €
Pack of 20 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
12902325
|
3M™
7000089667 |
4X-Large |
164.00 €
Pack of 20 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
11973734
|
3M™
7000089661 |
Small |
164.00 €
Pack of 20 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
Description
- Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts and limited liquid splashes
- Protection against EN 14126 Biological agents
- Made of a soft lightweight non-woven with microporous laminate for splash protection, strength and breathability
- Certified protection against Type 5/6 hazardous dusts and limited liquid splashes
- 2 panel hood design, 2-way zip with storm flap, elasticated apertures reduce the risk of skin exposure
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
Specifications
Cleanroom Coveralls | |
White | |
Elastic | |
2-way Zipper with Storm Flap | |
Non-sterile | |
EN 13034 Type 6, EN ISO 13982-1 Type 5, EN 1149-1:2006, ASTM D2859-96, CE | |
Abatement, Remediation, Agriculture, Cleaning and Maintenance, General Industry, Manufacturing, Non-Hazardous Operations, Painting, Pharmaceutical Cleanroom |
Microporous Polyethylene Laminate | |
Single-use | |
Elastic | |
Bound | |
Anti-static | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title