missing translation for 'onlineSavingsMsg'
Learn More

3M™ Disposable Protective Coveralls 4520, Hooded

Product Code. 12652595
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Coveralls protect against certain light liquid splashes and hazardous dusts while enhancing movement and comfort.

  • Designed for use in industrial workplaces as a physical barrier to help prevent skin contact from liquid splashes and dust in certain work environments
  • Seamless shoulder and sleeve tops help provide increased comfort and fewer potential entry points for contaminants
  • Meet ANSI 101-1996 size guidelines
  • Meets EN 340, Type 6 Splash - EN 13034/(EN ISO 17491-4), Type 5 Dust - EN ISO 13982-1, Anti-Static, EN 11492 and Nuclear - EN 1073 - 2
  • Microporous PE laminate
  • Breathable, SMS fabric back panel helps improve air ventilation and reduce heat build-up
  • Reinforcement gusset between the legs to help provide increased durability during bending and squatting
  • Elastic waist and ankles, knit cuffs, two-way zipper with sealable storm flap
  • Hooded style for use with respiratory equipment when extra protection is required
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Microporous Polyethylene Film
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Dust, Liquid
Coating Anti-static
Certifications/Compliance ANSI 101-1996
Certifications/Compliance (Europe) CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description 3 Layers of Melt-blown Polypropylene in-between 2 layers of Spunbound Polypropylene
Latex Free No
Model 4520
Size (English) Chest 45 to 49 in.
Size (English) Inseam 34 in.
Size (English) Waist 53 in.
Size (Metric) Chest 114.3 to 124.46 cm
Size (Metric) Inseam 86.36 cm
Size (Metric) Waist 134.62 cm
For Use With (Application) Insulation Installation, Asbestos and Lead Abatement, Coal Dust in Power Plants, Woodworking, Metal Polishing, Light-Duty Building Cleaning, General Powder Handling, General Industrial Clean-up and Maintenance, Machine or Vehicle Maintenance and Food Processing
Height (English) 6 to 6.33 ft.
Height (Metric) 186 to 194 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'