missing translation for 'onlineSavingsMsg'
Learn More

3M™ Disposable Protective Coveralls 4540+, Hooded

Product Code. 11943794
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Coveralls are designed to help protect against hazardous dusts (Type 5) and certain light liquid splashes (Type 6).

  • Premium dual material technology provides both protective performance and comfort
  • Barrier against dry particles and limited liquid chemical splash (CE Type 5/6)
  • Soft and light, super low-linting laminated microporous material
  • Back panel made of breathable SMS material for improved air circulation and reduced heat stress
  • Two-way zipper with fully sealable storm flap
  • Three-panel hood design for enhanced compatability with complimentary PPE
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
  • Anti-static treated
  • Low linting
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Polyethylene, Polyester/Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged, Bound
Sterility Non-sterile
Resistance Anti-static, Dust, Liquid
Coating Anti-static
Back Style Breathable SMMMS material
Certifications/Compliance ANSI 101-1996
Certifications/Compliance (Europe) CE Category III Type 5/6, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description White Polypropylene/Polyethylene laminated suit, SMMMS Polypropylene blue back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread
Latex Free No
Model 4540+
Size (English) Chest 52 to 55 in.
Size (English) Inseam 36 in.
Size (English) Waist 55 in.
Size (Metric) Chest 132.08 to 139.7 cm
Size (Metric) Inseam 91.44 cm
Size (Metric) Waist 139.7 cm
For Use With (Application) Pesticide, Asbestos and Lead Abatement, Power Plants, Metals, Cleaning, Maintenance, General Industrial, Insulation, Food Processing, Pharmaceutical
Height (English) 6.5 to 6.75 ft.
Height (Metric) 200 to 206 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'