Learn More
3M™ Accessories and Replacement Parts for 6000 Series Full Facepieces, Nose Cup Assembly
For use as a replacement part for 3M™ Full Facepieces 6700, 6800, 6900 (07138, 07139, 07140 AAD).
Brand: 3M™ 7000002034
23.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory to shield eyes from bright, intense light (not for welding)
- Tinting relaxes eyes to make it easier to focus on outdoor work
- Designed for use with 3M Full Facepiece 6000 Series
- Cover fits easily over lens
- Approved 3M protection accessory
- 3M Catalog# 6894
Specifications
Nosecup Assembly | |
Grinding, welding | |
Full facepiece 6700, 6800, 6900 | |
6894 |