missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 3M™ Comply™ Indicator Tape

Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.

Brand:  Thermo Scientific™ 7000138230

 View more versions of this product

Product Code. 11761351

  • 6.90 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.

As an exposure monitor, 3M™ indicator tapes securely seal packs and enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilization process. The chemical indicator ink, printed in a diagonal striped pattern, changes colour or turns darker when exposed to the sterilisation process.
  • Lead-free steam indicator tape is designed for use on all wraps
  • Reliable achesive adheres to packaging material
  • Sustainable, solvent-free manufacturing proces
  • Does not require hazardous waste disposal
  • Redesigned packaging uses up to 30% less plastic
  • Convenient, individually wrapped, easy-to-open rolls
  • Lead free and no added latex
Specifications

Specifications

Meets NF EN 867-1 Class A, ISO 11140-1 Class 1
55 m x 24 mm
Autoclave
Indicator Tape
Yes
24 mm
Beige
Sterilization verification
55 m
Lead-free, aggressive, pressure sensitive adhesive
Steam
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers