missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 3M™ Comply™ Indicator Tape

Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.

Brand:  Thermo Scientific™ 7000138229

 View more versions of this product

Product Code. 12986976

  • 9.05 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.

As an exposure monitor, 3M™ indicator tapes securely seal packs and enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. The chemical indicator ink, printed in a diagonal striped pattern, changes colour or turns darker when exposed to the sterilisation process.
  • Lead-free steam indicator tape is designed for use on all wraps
  • Reliable achesive adheres to packaging material
  • Sustainable, solvent-free manufacturing proces
  • Does not require hazardous waste disposal
  • Redesigned packaging uses up to 30% less plastic
  • Convenient, individually wrapped, easy-to-open rolls
  • Lead free and no added latex
Specifications

Specifications

Meets NF EN 867-1 Class A, ISO 11140-1 Class 1
55 m x 18 mm
Autoclave
Indicator Tape
Each
Steam
Beige
Sterilization verification
55 m
Lead-free, aggressive, pressure sensitive adhesive
Yes
18 mm
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 3M™ Comply™ Indicator Tape > Width: 18mm; Material: Adhesive Paper

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.