missing translation for 'onlineSavingsMsg'
Learn More

3M™ Disposable Protective Lab Coat, Series 4440

Product Code. 15805121
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Offers a barrier against non-hazardous particles and liquid splashes in light duty applications.

  • Breathable, lightweight material.
  • Designed for light duty applications.
  • Offers a barrier against non-hazardous particles and liquid splashes.
  • Zip fastener.
  • Anti-static treated.
  • Knitted cuffs.
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Zipper
Cuff Style Knit
Disposable Single-use
Model 4440-2XL
Seam Type Serged
Size (English) Chest 40 to 43 in.
Size (Metric) Chest 102 to 109 cm
Certifications/Compliance (Europe) CE Category 1
For Use With (Application) Catering, Cleanrooms, Electronics Manufacturing, General Maintenance, Laboratories
Weight (English) 1.93 oz.
Construction Microporous Laminate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'