missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Ethylene Oxide Monitor
No batteries, hoses, pumps or equipment to calibrate
Brand: 3M™ 7000001562
This item is not returnable.
View return policy
Description
- Ideal for chemical, medical, and pharmaceutical labs.
- Sensitivity of 0.24ppm-hours
- Upper exposure limit of 600ppm-hours
- Available as a monitor only or with prepaid analysis
- These safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
- 3M Catalog# 3551
Compliance
Meets OSHA accuracy requirements.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Ideal for chemical, medical, and pharmaceutical labs | |
Ethylene Oxide | |
Ethylene Oxide Monitor with Prepaid analysis |
OSHA | |
600 ppm.Hour |