Learn More
3M™ PELTOR™ Hygiene Kit for X Series Ear Muffs
missing translation for 'orderingAttributeHoverText'
Model:
HYX1
HYX2
HYX3
HYX4
HYX5
For Use With (Equipment):
PELTOR™ X1 Ear Muffs
PELTOR™ X2 Ear Muffs
PELTOR™ X3 Ear Muffs
PELTOR™ X4 Ear Muffs
PELTOR™ X5 Ear Muffs
Description
- Hygiene kits consist of one pair of replacement cushions and foam inserts for 3M™ PELTOR™ X Series ear muffs (X1, X2, X3, X4, X5)
- 3M recommends replacing ear muff cushions and foam inserts every six months or whenever they become damaged.
- Kits are packaged 10 Each/Cs.
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Spécification
Spécification
Cushion Color | Black |
For Use With (Equipment) | PELTOR™ X4 Ear Muffs |
For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding |
Model | HYX4 |
Product Type | Hygiene Kit |
Dielectric | Yes |