Learn More
3M™ Standard Chemical Sorbents
Absorb virtually any liquid with these chemical sorbents
Brand:  3M™ P-500
 View more versions of this product
Product Code. 12362459
Description
Description
Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.
  • Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
  • Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
  • Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
  • Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
  • Mini booms surround and contain spills; pillows soak them up
  • 3M Catalog# P110, P200, P208, P300, P500
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OSHA 29 CFR 1910.132
14 gal.
Polypropylene
For control of spills of certain hazardous liquids, particulate materials are used in packaging applications where breakage of a container could cause a release of a hazardous liquid
Yellow
53 L
Chemical Sorbent Particulate
P500
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Standard Chemical Sorbents > Bag; 12 lb. (5.4kg); Sorbing Capacity: 14 gal. (53L)