missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Head, Neck and Shoulder Cover for M-100 and M-300 Series PAPR

Product Code. 15770078
Click to view available options
:
Head, neck and shoulder cover
This item is not returnable. View return policy

Product Code. 15770078

missing translation for 'mfr': 3M™ 7100063944

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with M-100 and M-300 Series PAPR

  • Material: Single Layer Polypropylene
  • 3M Catalog# M-976

TRUSTED_SUSTAINABILITY

Specifications

Color White
Disposable Reusable
Material Polypropylene
Product Type Head, Neck, Shoulder Cover
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo™ M-100 and M-300 Series
Model M-976
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.