missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ Heavy Duty Neoprene Rubber Breathing Tube
Breathing tube made of heat-resistant neoprene rubber material for rugged applications.
Brand: 3M™ 7100069524
123.75 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- High durability air breathing tube that is easy to wipe down
- Quick release swivel enables one-handed connection and release.
- Compatible with all 3M™ Versaflo™ Headtops and Air Delivery Units
- Size: 840 mm
- 3M Catalog# BT-40
Specifications
Neoprene | |
Assembly, facility maintenance, grinding, machine operation, painting, sanding | |
BT-40 |
Breathing Tube | |
Versaflo™ TR-300 Series PAPR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction