missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ M-300 Series Respiratory Hardhat Assembly
Versaflo™ M-300 Series Respiratory Hardhat Assembly provides respiratory and limited head and eye protection.
600.00 €
Specifications
Assigned Protection Factor (APF) | 25 |
---|---|
Certifications/Compliance | CE Approved, EN 12941 TH2/TH3, EN 14594 3B, EN 166:2001, EN 397 |
Color | Black |
Product Type | Respirator Hard Hat |
For Use With (Application) | Demolition, Food and Beverage Manufacturing, Metalworking, Pulp and Paper, Agriculture, Smelting, Woodworking, Chemical Processing, Construction |
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
11773916
|
3M™
7000104036 |
M-307 | Flame-Resistant Faceseal | ||||||
Description
Features:
- Chemical and Scratch Resistant Lenses
- Large Viewing Area
- Lightweight, Easy Visor Replacement System
Environmental Conditions:
- Cold Working Environment
- High Mobility Required
- Safety Helmet Required
Hazard Type:
- Gas, Vapour and Particulates
- Liquid Splash
- Low Energy Flying Particles/Dust
- Falling Objects
- Sparks and Hot Particles
Applications:
- Automotive, food and beverage manufacturing
- Metal production and fabrication
- Military maintenance
- Repair and operation
- Pharmaceutical
- Transportation
Specifications
25 | |
Black | |
Demolition, Food and Beverage Manufacturing, Metalworking, Pulp and Paper, Agriculture, Smelting, Woodworking, Chemical Processing, Construction | |
Non-sterile | |
Yes | |
13.6 in. | |
13.6 in. | |
9.6 in. | |
1.8 lb. |
CE Approved, EN 12941 TH2/TH3, EN 14594 3B, EN 166:2001, EN 397 | |
Respirator Hard Hat | |
3M Air Delivery Units | |
Textile | |
346 x 346 x 254 mm | |
346 mm | |
346 mm | |
254 mm | |
832 g |