Learn More
3M™ Versaflo™ TR-600 Series PAPR Accessory, Air Flow Indicator TR-971
Enables confirmation of airflow levels of TR-600 series PAPR.
Brand: 3M™ 7100035846
26.15 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Confirms factory calibration of airflow
- Designed to enable testing at various elevations and temperatures within operational range of PAPR system
- Easy to use
- 3M Catalog# TR-971
Applications:
Recommended Industry: Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Certifications
NIOSH Approved
Specifications
Reusable | |
Versaflo™ TR-600 Series PAPR System | |
1 Ea. |
Airflow Indicator | |
TR-971 |