missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ TR-600 Series PAPR Belts and Extenders

For use with Versaflo™ TR-600 Series PAPR

Brand:  3M™ 7100222675

 View more versions of this product

Product Code. 15569255

  • 141.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TR-626 High Durability Belt

  • Comfortable belt due to ergonomic design
  • Durable construction and easy to use
  • Leather leads make it suitable for grinding
  • Part of TR-613N and TR-616N assemblies, TR-600-HIK Ready-to-Use Kit
  • Accommodates waist sizes up to 52 in.
  • Applications: Healthcare, Shutdown, Pharmaceutical, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
  • Color: Gray
  • TR-626X Belt Extender
    • Comfortable due to ergonomic design
    • Easy to use extender
    • Enables TR-626 belt to be worn comfortably, when additional belt length is needed
    • Multiple TR-626X Belt Extenders may be connected in series
    • Applications: Healthcare, Shutdown, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
    • Color: Gray
    • TR-627 Easy Clean Belt
      • Comfortable due to ergonomic design
      • Easy to use belt
      • Smooth surfaces make it easier to clean
      • Part of TR-614N and TR-617N assemblies, TR-600-ECK Ready-to-Use Kit
      • Accommodates waist sizes up to 52 in.
      • Applications: Healthcare, Pharmaceutical, Pesticide Application, Animal processing, Laboratories, Nuclear energy, Cosmetics, Pharmaceutical research and production, Recycling, Dry Foods
      • Color: Gray
      • TR-627X Belt Extender
        • Comfortable due to ergonomic design
        • Easy to use extender
        • Enables TR-627 belt to be worn comfortably, when additional belt length is needed
        • Multiple TR-627X Belt Extenders may be connected in series
        • Applications: Healthcare, Pharmaceutical, Pesticide Application, Animal processing, Laboratories, Nuclear energy, Cosmetics, Pharmaceutical research and production, Recycling, Dry Foods
        • Color: Gray
        • 3M Catalog# TR-626, TR-626X, TR-627, TR-627X

        Certifications

        NIOSH Approved

        TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gray
Belt
Versaflo™ TR-600 Series PAPR
1 Ea.
Reusable
Healthcare, Shutdown, Pharmaceutical, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
TR-626
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.