Learn More
3M™ Versaflo™ TR-Series PAPR Accessory, Spark Arrestor
When installed inside the filter cover of the TR-300 PAPR, the TR-362 spark arrestor helps protect the HE filter and PAPR unit from damage due to sparks or other hot materials.
Brand:  3M™ 7000103970
9.89 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 15709897
  • 13.00 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 3M Catalog# TR-362
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reusable
Protects The he Filter and Powered Air Purifying Respirator Unit From Damage Due to Sparks or Other Hot Materials
TR-362
Spark Arrestor
Versaflo TR-Series PAPR System
2/Cs.