missing translation for 'onlineSavingsMsg'
Learn More

(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10490311
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4272-74-6
Molecular Formula C14H22Cl2N2O3S
Molecular Weight (g/mol) 369.30
MDL Number MFCD00065395
InChI Key YFCUZWYIPBUQBD-ZOWNYOTGSA-N
Synonym tlck hydrochloride, tlck, s-n-7-amino-1-chloro-2-oxoheptan-3-yl-4-methylbenzenesulfonamide hydrochloride, 3s-1-chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, n-alpha-p-tosyl-l-lysine chloromethyl ketone hydrochloride, na-p-tosyl-l-lysine chloromethyl ketone hydrochloride, c14h21cln2o3s hydrochloride, tosyl-l-lysyl-chloromethane hydrochloride, l-n-5-amino-1-chloroacetyl pentyl-p-toluenesulfonamide hydrochloride, p-toluenesulfonamide, n-5-amino-1-chloroacetyl pentyl-, hydrochloride, l
PubChem CID 73093
IUPAC Name N-[(3S)-7-amino-1-chloro-2-oxoheptan-3-yl]-4-methylbenzenesulfonamide;hydrochloride
SMILES [H+].[Cl-].CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160°C to 161°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2N(CH2)4CH(NHSO2C6H4CH3)COCH2Cl·HCl
Quantity 50 mg
Specific Rotation -9.5
Formula Weight 369.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'