Learn More
UVP 3UV™ Lamp
Innovative, versatile, and patented UV lamp for multiple applications. UVP™ 3UV™ Lamp provides longwave, shortwave and midrange UV light in one lamp.
Brand:  UVP 95-0343-02
Product Code. 11743539
  • 1790.00 € / Each
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Three wavelengths, 254/302/365nm UV, in one lamp
  • Selection of each wavelength is by an easy turn of the dial located on one end of the housing
  • The uniquely designed reflector surfaces mounted behind each tube provides maximum UV for fluorescence applications
  • Lightweight and ergonomically designed for hand or stationary use
  • Design allows the stationary use by standing the lamp on its end or side
  • Available in four, six or eight watts
  • Applications:
    • Inspection
    • Laboratory
    • Education
Specifications
Specifications
230 V
365/302/254 nm
8 W, 230 V, 50 Hz
3UV-38
8 W
394 x 76 x 114 mm
50 Hz
3UV Lamp
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
UVP 3UV™ Lamp > Model: 3UV-38