missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Dioxolan-2-yl)benzene-1-carbothioamide, 97%, Thermo Scientific™

Product Code. 10049361
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 175202-43-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H11NO2S
MDL Number MFCD00085174
Quantity 1 g
Synonym 4-1,3-dioxolan-2-yl benzene-1-carbothioamide, 4-1,3-dioxolan-2-yl benzenecarbothioamide, benzenecarbothioamide,4-1,3-dioxolan-2-yl, maybridge1_004514, 4-1,3-dioxolan-2-yl thiobenzamide, 4-1,3-dioxolan-2-yl benzothioamide, amino 4-1,3-dioxolan-2-yl phenyl methane-1-thione
InChI Key HIAHUZPFVWVKOR-UHFFFAOYSA-N
SMILES C1COC(O1)C2=CC=C(C=C2)C(=S)N
IUPAC Name 4-(1,3-dioxolan-2-yl)benzenecarbothioamide
Molecular Weight (g/mol) 209.263
PubChem CID 2778731
Formula Weight 209.27
Percent Purity 97%
Chemical Name or Material 4-(1, 3-Dioxolan-2-yl)benzene-1-carbothioamide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(1,3-Dioxolan-2-yl)benzene-1-carbothioamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.