missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™

Product Code. 10177552
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 112809-25-3
Molecular Formula C10H8N4
Molecular Weight (g/mol) 184.202
MDL Number MFCD07368016
InChI Key HQLYWHSJALKYOV-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl benzonitrile, 4-1h-1,2,4-triazol-1-yl methyl benzonitrile, 4-1h-1,2,4-triazolylmethyl benzonitrile, 4-1,2,4-triazol-1-ylmethyl benzonitrile, benzonitrile, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4-triazol-1-yl methyl benzonitrile, 4-1-1,2,4-triazolyl methyl benzontrile, 4-1,2,4-triazolylmethyl benzenecarbonitrile, letrozole impurity c
PubChem CID 7537616
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)benzonitrile
SMILES C1=CC(=CC=C1CN2C=NC=N2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 184.2
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-triazol-1-ylmethyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'