missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-1,2,4-triazol-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 25 g |
Formula Weight | 184.2 |
Percent Purity | 97% |
Chemical Name or Material | 4-(1H-1, 2, 4-triazol-1-ylmethyl)benzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'