missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Pyrazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 4-1h-pyrazol-1-ylmethyl benzoic acid, 4-pyrazol-1-ylmethyl-benzoic acid, 4-pyrazol-1-ylmethyl benzoic acid, benzoicacid, 4-1h-pyrazol-1-ylmethyl, 4-pyrazolylmethyl benzoic acid, 4-1h-pyrazol-1-yl methyl benzoic acid, 4-1h-pyrazole-1-ylmethyl benzoic acid, benzoicacid,4-1h-pyrazol-1-ylmethyl, benzoic acid, 4-1h-pyrazol-1-ylmethyl |
Formula Weight | 202.21 |
Percent Purity | 97% |
Chemical Name or Material | 4-(1H-Pyrazol-1-ylmethyl)benzoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction