missing translation for 'onlineSavingsMsg'
Learn More

[4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol 97+%, Thermo Scientific™

Product Code. 11306805
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 160388-55-6
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD08435916
InChI Key ADLULGOPZPFWPA-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-ylmethyl phenyl methanol, 4-pyrazol-1-ylmethyl phenyl methanol, 4-1h-pyrazol-1-yl methyl phenyl methanol, 1-4-hydroxymethyl-benzyl-1h-pyrazole, benzenemethanol, 4-1h-pyrazol-1-ylmethyl, 9vq
PubChem CID 16640562
IUPAC Name [4-(pyrazol-1-ylmethyl)phenyl]methanol
SMILES OCC1=CC=C(CN2C=CC=N2)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material [4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.