missing translation for 'onlineSavingsMsg'
Learn More
4-(1H-Pyrrol-1-yl)benzonitrile, 97%, Thermo Scientific™ Product Code.: 10669515

4-(1H-Pyrrol-1-yl)benzonitrile, 97%, Thermo Scientific™

Product Code. 10669515
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 10669515

Brand: Thermo Scientific Maybridge CC25516EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 23351-07-7
Molecular Formula C11H8N2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00085164
InChI Key OKVSZRKKRHNDOL-UHFFFAOYSA-N
Synonym 4-1h-pyrrol-1-yl benzonitrile, 1-4-cyanophenyl pyrrole, n-4-cyanophenyl pyrrole, benzonitrile, 4-pyrrol-1-yl, 4-1-pyrrolyl benzonitrile, 4-pyrrol-1-yl benzonitrile, 4-pyrrolylbenzenecarbonitrile, acmc-1cekp, maybridge1_001756, 1-4-cyanophenyl-pyrroleShow MoreShow Less
PubChem CID 272424
SMILES N#CC1=CC=C(C=C1)N1C=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
IUPAC Name 4-pyrrol-1-ylbenzonitrile
Formula Weight 168.2
Percent Purity 97%
Chemical Name or Material 4-(1H-pyrrol-1-yl)benzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(1H-Pyrrol-1-yl)benzonitrile, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.