Learn More
4-(2-Aminoethyl)pyridine, 97%, Thermo Scientific Chemicals
162.81 € - 565.82 €
Chemical Identifiers
CAS | 13258-63-4 |
---|---|
Molecular Formula | C7H10N2 |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00038045 |
InChI Key | IDLHTECVNDEOIY-UHFFFAOYSA-N |
Synonym | 4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine |
PubChem CID | 83275 |
IUPAC Name | 2-pyridin-4-ylethanamine |
SMILES | NCCC1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11401840
|
Thermo Scientific Alfa Aesar
H27545.03 |
1 g |
162.81 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11411840
|
Thermo Scientific Alfa Aesar
H27545.06 |
5 g |
565.82 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13258-63-4 | |
122.17 | |
IDLHTECVNDEOIY-UHFFFAOYSA-N | |
83275 | |
NCCC1=CC=NC=C1 |
C7H10N2 | |
MFCD00038045 | |
4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine | |
2-pyridin-4-ylethanamine |
Specifications
13258-63-4 | |
70°C to 71°C (1 mmHg) | |
Amine-like | |
1.5395 | |
UN2735 | |
4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine | |
NCCC1=CC=NC=C1 | |
122.17 | |
122.17 | |
4-(2-Aminoethyl)pyridine |
1.021 | |
>110°C (230°F) | |
C7H10N2 | |
MFCD00038045 | |
Air Sensitive | |
IDLHTECVNDEOIY-UHFFFAOYSA-N | |
2-pyridin-4-ylethanamine | |
83275 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.