missing translation for 'onlineSavingsMsg'
Learn More

4-{[2-(Bromomethyl)phenyl]sulfonyl}morpholine, 90%, Thermo Scientific™

Product Code. 10451884
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 941717-06-2
Molecular Formula C11H14BrNO3S
Molecular Weight (g/mol) 320.201
MDL Number MFCD09879984
InChI Key VRRKTYLWVKMVJU-UHFFFAOYSA-N
Synonym 4-2-bromomethyl phenyl sulfonyl morpholine, 2-morpholin-4-yl sulphonyl benzyl bromide, 4-2-bromomethyl benzenesulfonyl morpholine, 4-2-bromomethyl phenyl sulphonyl morpholine, 2-morpholin-4-yl sulfonyl benzyl bromide, 4-2-bromomethyl benzene-1-sulfonyl morpholine
PubChem CID 24229771
IUPAC Name 4-[2-(bromomethyl)phenyl]sulfonylmorpholine
SMILES C1COCCN1S(=O)(=O)C2=CC=CC=C2CBr

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 320.21
Percent Purity 90%
Chemical Name or Material 4-{[2-(Bromomethyl)phenyl]sulfonyl}morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'