missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethyl)phenol, 98%, Thermo Scientific Chemicals

Product Code. 11475567
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56718-71-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00017537
InChI Key FAYGEALAEQKPDI-UHFFFAOYSA-N
Synonym 4-2-methoxyethyl phenol, p-2-methoxyethyl phenol, phenol, 4-2-methoxyethyl, 4-methoxyethylphenol, unii-py1w0akw8i, metoprolol impurity b, metoprolol impurity i, py1w0akw8i, 4-hydroxyphenylethyl methyl ether
PubChem CID 92516
IUPAC Name 4-(2-methoxyethyl)phenol
SMILES COCCC1=CC=C(C=C1)O

Specifications

Melting Point 43°C to 45°C
Quantity 50 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-(2-Methoxyethyl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'