missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methylphenoxy)piperidine, 97%, Thermo Scientific Chemicals

76.60 € - 280.00 €

Chemical Identifiers

CAS 63843-42-5
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD06248344
InChI Key MLLZZLRYRWFFRQ-UHFFFAOYSA-N
Synonym 4-o-tolyloxy piperidine, 4-2-methylphenoxy piperidine, 4-o-tolyloxy-piperidine, acmc-1b7xi, piperidine,4-2-methylphenoxy
PubChem CID 3686170
IUPAC Name 4-(2-methylphenoxy)piperidine
SMILES CC1=CC=CC=C1OC2CCNCC2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11419700
View Documents
Thermo Scientific Alfa Aesar
H50951.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11429700
View Documents
Thermo Scientific Alfa Aesar
H50951.03
1 g
280.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63843-42-5
191.274
MLLZZLRYRWFFRQ-UHFFFAOYSA-N
3686170
CC1=CC=CC=C1OC2CCNCC2
C12H17NO
MFCD06248344
4-o-tolyloxy piperidine, 4-2-methylphenoxy piperidine, 4-o-tolyloxy-piperidine, acmc-1b7xi, piperidine,4-2-methylphenoxy
4-(2-methylphenoxy)piperidine

Specifications

63843-42-5
MFCD06248344
4-o-tolyloxy piperidine, 4-2-methylphenoxy piperidine, 4-o-tolyloxy-piperidine, acmc-1b7xi, piperidine,4-2-methylphenoxy
CC1=CC=CC=C1OC2CCNCC2
191.274
191.28
4-(2-Methylphenoxy)piperidine
C12H17NO
Air Sensitive
MLLZZLRYRWFFRQ-UHFFFAOYSA-N
4-(2-methylphenoxy)piperidine
3686170
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only