missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ 4 – 20 mA Current Loop Interface CO2 Controller
4-20mA current loop interface CO2 controller 0-25%
244.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17100045
|
Memmert™
ICO50V9 |
ICO50 |
244.00 €
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
17109944
|
Memmert™
ICO105V9 |
ICO105 |
244.00 €
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
17139974
|
Memmert™
ICO150V9 |
ICO150 |
244.00 €
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
17170005
|
Memmert™
ICO240V9 |
ICO240 |
244.00 €
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
Description
- Accessory
- Actual value (0 – 25 % CO2 4 – 20 mA)
Specifications
CO2 Controller |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title