Learn More
Memmert™ 4-20 mA Current Loop Interface for UF30, Actual Value
Product Code. 17569315 Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Product Code. 17569315
Brand: Memmert™ UF30V3
Only null left
Add to basket
Add to basket
Temperature Output for external temperature monitoring.
  • Temperature controller actual value
Specifications
For Use With (Equipment) UF30 Oven
Type Current Loop Interface
Amperage 4 to 20 mA