missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]benzonitrile, 97%, Thermo Scientific™

Product Code. 10356994
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 146440-15-5
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD09064993
InChI Key XYCJVOUBTBJYTL-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy benzonitrile, benzonitrile,4-3-dimethylamino propoxy, acmc-20n4ty, benzonitrile, 4-3-dimethylamino propoxy, 4-3-dimethylamino propoxy benzenecarbonitrile
PubChem CID 11858362
IUPAC Name 4-[3-(dimethylamino)propoxy]benzonitrile
SMILES CN(C)CCCOC1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 4-[3-(Dimethylamino)propoxy]benzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.