missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde, 90%, Thermo Scientific™

Product Code. 10245964
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-27-7
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD09025865
InChI Key AEMXYVCSLPLJOW-UHFFFAOYSA-N
Synonym 4-3-methyl-1,2,4-oxadiazol-5-yl benzaldehyde, 3-methyl-5-4-formylphenyl-1,2,4-oxadiazole, benzaldehyde,4-3-methyl-1,2,4-oxadiazol-5-yl
PubChem CID 18525855
IUPAC Name 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
SMILES CC1=NOC(=N1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.19
Percent Purity 90%
Chemical Name or Material 4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'