missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)-1H-indole, ≥97%, Thermo Scientific™

Product Code. 10601675
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-17-8
Molecular Formula C23H38BNO2Si
Molecular Weight (g/mol) 399.46
MDL Number MFCD05865142
InChI Key NGMKDRPSWHSLIM-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl-1h-indole, 1-tris isopropylsilyl-1h-indole-4-boronic acid, pinacol ester, 1-triisopropylsilyl-1h-indole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris isopropylsilyl-1h-indole, 1-triisopropylsilyl-1h-indole-4-boronic acid,pinacol ester, 1-triisopropylsilyl indole-4-boronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl indole, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-triisopropylsilanyl-1h-indole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tri propan-2-yl silyl-1h-indole, tri propan-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indol-1-yl silane
PubChem CID 2795574
IUPAC Name tri(propan-2-yl)-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indol-1-yl]silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 399.46
Percent Purity 97%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)-1H-indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'