missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Trimethyltrityl alcohol, 98+%, Thermo Scientific Chemicals

Product Code. 11387685
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3247-00-5
Molecular Formula C22H22O
Molecular Weight (g/mol) 302.417
MDL Number MFCD00014919
InChI Key DNWQXZDDISHGRM-UHFFFAOYSA-N
Synonym 4,4',4-trimethyltrityl alcohol, tris 4-methylphenyl methanol, methanol, tri-p-tolyl, tri-p-tolylmethanol, tri p-tolyl methanol, 4,4'-dimethyl-4-methyltrityl alcohol, benzenemethanol, 4-methyl-.alpha.,.alpha.-bis 4-methylphenyl, acmc-20amt7, tris 4-methylphenyl methan-1-ol, 4,4-trimethyltriphenylmethanol
PubChem CID 76733
IUPAC Name tris(4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)(C3=CC=C(C=C3)C)O

Specifications

Melting Point 92°C to 96°C
Quantity 5 g
Beilstein 2055803
Formula Weight 302.42
Percent Purity ≥98%
Chemical Name or Material 4,4',4''-Trimethyltrityl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'