Learn More
4,4',4″-Tris(carbazol-9-yl)triphenylamine, 99%, Thermo Scientific Chemicals
70.45 € - 220.10 €
Chemical Identifiers
CAS | 139092-78-7 |
---|---|
Molecular Formula | C54H36N4 |
Molecular Weight (g/mol) | 740.91 |
MDL Number | MFCD03093250 |
InChI Key | AWXGSYPUMWKTBR-UHFFFAOYSA-N |
Synonym | tris 4-9h-carbazol-9-yl phenyl amine, tcta, tcata, 4,4',4-tris carbazol-9-yl-triphenylamine, 4,4',4-tri-9-carbazolyltriphenylamine, tris 4-carbazoyl-9-ylphenyl amine, tris 4-carbazol-9-ylphenyl amine, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-yl phenyl aniline, 4,4',4-tris carbaz, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-ylphenyl aniline |
PubChem CID | 9962045 |
IUPAC Name | 4-carbazol-9-yl-N,N-bis(4-carbazol-9-ylphenyl)aniline |
SMILES | C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15575996
|
Thermo Scientific Acros
459180010 |
1 g |
70.45 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15585996
|
Thermo Scientific Acros
459180050 |
5 g |
220.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
139092-78-7 | |
740.91 | |
AWXGSYPUMWKTBR-UHFFFAOYSA-N | |
9962045 | |
C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
C54H36N4 | |
MFCD03093250 | |
tris 4-9h-carbazol-9-yl phenyl amine, tcta, tcata, 4,4',4-tris carbazol-9-yl-triphenylamine, 4,4',4-tri-9-carbazolyltriphenylamine, tris 4-carbazoyl-9-ylphenyl amine, tris 4-carbazol-9-ylphenyl amine, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-yl phenyl aniline, 4,4',4-tris carbaz, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-ylphenyl aniline | |
4-carbazol-9-yl-N,N-bis(4-carbazol-9-ylphenyl)aniline |
Specifications
139092-78-7 | |
86.23 to 88.85% Carbon 7.45 to 7.68% Nitrogen | |
99% | |
C54H36N4 | |
tris 4-9h-carbazol-9-yl phenyl amine, tcta, tcata, 4,4',4-tris carbazol-9-yl-triphenylamine, 4,4',4-tri-9-carbazolyltriphenylamine, tris 4-carbazoyl-9-ylphenyl amine, tris 4-carbazol-9-ylphenyl amine, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-yl phenyl aniline, 4,4',4-tris carbaz, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-ylphenyl aniline | |
C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
740.91 | |
740.92 | |
4,4',4''-Tris(carbazol-9-yl)triphenylamine |
297.0°C to 300.0°C | |
Authentic | |
Glass Bottle | |
MFCD03093250 | |
AWXGSYPUMWKTBR-UHFFFAOYSA-N | |
4-carbazol-9-yl-N,N-bis(4-carbazol-9-ylphenyl)aniline | |
9962045 | |
99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
GHS Signal Word: Warning
- 4,4',4"-Tris(carbazol-9-yl)triphenylamine, 99%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.