missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[4-(thien-2-ylmethyl)phenyl]-1,3,2-dioxaborolane, 97%, Thermo Scientific™

Product Code. 10707695
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1007847-76-8
Molecular Formula C17H21BO2S
Molecular Weight (g/mol) 300.22
MDL Number MFCD11841077
InChI Key OPJUSDABOPHYPQ-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-4-thien-2-ylmethyl phenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-thiophen-2-ylmethyl phenyl-1,3,2-dioxaborolane, 4-thien-2-ylmethyl benzeneboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-thiophen-2-yl methyl phenyl-1,3,2-dioxaborolane
PubChem CID 43811052
IUPAC Name 4,4,5,5-tetramethyl-2-[4-(thiophen-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CC2=CC=CS2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 300.23
Percent Purity 97%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[4-(thien-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'