missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Acetyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 15445256
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218791-38-8
Molecular Formula C18H27BN2O3
Molecular Weight (g/mol) 330.235
MDL Number MFCD13195756
InChI Key VRZVSHHLWMAHAZ-UHFFFAOYSA-N
Synonym 4-4-acetyl-1-piperazinyl phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethanone, 4-4-acetylpiperazino phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1-piperazinyl ethanone, 4-4-acetyl-1-piperazinyl phenylboronicacidpinacolester, 4-4-acetylpiperazino phenylboronic acid, pinacol ester, 4-4-acetyl-1-piperazinyl benzeneboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethan-1-one
PubChem CID 53217141
IUPAC Name 1-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl]ethanone
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(=O)C

Specifications

Quantity 250 mg
Formula Weight 330.23
Percent Purity 95%
Chemical Name or Material 4-(4-Acetyl-1-piperazinyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'