missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(dimethylamino)benzil, 98%, Thermo Scientific Chemicals

Product Code. 11975321
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17078-27-2
Molecular Formula C18H20N2O2
Molecular Weight (g/mol) 296.36
InChI Key AVFUVYIDYFXFSX-UHFFFAOYSA-N
Synonym 4,4'-bis dimethylamino benzil, 1,2-bis 4-dimethylamino phenyl ethane-1,2-dione, ethandione, bis 4-dimethylamino phenyl, bis 4-dimethylamino phenyl ethane-1,2-dione, acmc-209e1j, benzil-based compound, 35, bis n,n-dimethylamino benzil, 4,4/'-bis dimethylamino benzil, bis 4-dimethylamino phenyl diketone
PubChem CID 222570
IUPAC Name 1,2-bis[4-(dimethylamino)phenyl]ethane-1,2-dione
SMILES CN(C)C1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)N(C)C

Specifications

Melting Point 200.0°C to 204.0°C
Color Green to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 296.36
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Bis(dimethylamino)benzil
Product Identifier
  • 4,4'-Bis(dimethylamino)benzil

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.