missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

76.00 € - 294.00 €

Chemical Identifiers

CAS 936694-19-8
Molecular Formula C22H35BN2O4
Molecular Weight (g/mol) 402.342
MDL Number MFCD16294502
InChI Key AAEYFMAHSYKHGD-UHFFFAOYSA-N
Synonym tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, 4-4-boc-piperazinomethyl phenylboronic acid, pinacol ester, 4-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, amtb122, 4-4-boc-piperazinemethyl phenylboronic acid pinacol ester, 4-4-boc-piperazine methyl phenylboronic acid pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzyl piperazine-1-carboxylate
PubChem CID 53216820
IUPAC Name tert-butyl 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3CCN(CC3)C(=O)OC(C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15439315
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51743.03
1 g
76.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
15449315
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51743.06
5 g
294.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

936694-19-8
402.342
AAEYFMAHSYKHGD-UHFFFAOYSA-N
53216820
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3CCN(CC3)C(=O)OC(C)(C)C
C22H35BN2O4
MFCD16294502
tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, 4-4-boc-piperazinomethyl phenylboronic acid, pinacol ester, 4-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, amtb122, 4-4-boc-piperazinemethyl phenylboronic acid pinacol ester, 4-4-boc-piperazine methyl phenylboronic acid pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzyl piperazine-1-carboxylate
tert-butyl 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate

Specifications

936694-19-8
MFCD16294502
tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, 4-4-boc-piperazinomethyl phenylboronic acid, pinacol ester, 4-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, amtb122, 4-4-boc-piperazinemethyl phenylboronic acid pinacol ester, 4-4-boc-piperazine methyl phenylboronic acid pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzyl piperazine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3CCN(CC3)C(=O)OC(C)(C)C
402.342
402.34
4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
C22H35BN2O4
18669672
AAEYFMAHSYKHGD-UHFFFAOYSA-N
tert-butyl 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate
53216820
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only