missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-bromothien-2-yl)methyl]morpholine, 97%, Thermo Scientific™

Product Code. 10525944
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 194851-19-9
Molecular Formula C9H12BrNOS
Molecular Weight (g/mol) 262.17
MDL Number MFCD01653482
InChI Key XYQVGDMXJNORKK-UHFFFAOYSA-N
Synonym 4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl
PubChem CID 783497
IUPAC Name 4-[(4-bromothiophen-2-yl)methyl]morpholine
SMILES BrC1=CSC(CN2CCOCC2)=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 262.17
Percent Purity 97%
Chemical Name or Material 4-[(4-Bromothien-2-yl)methyl]morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'