missing translation for 'onlineSavingsMsg'
Learn More

4,4″-Diamino-p-terphenyl, 95%, Thermo Scientific Chemicals

Product Code. 11397425
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3365-85-3
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00051532
InChI Key QBSMHWVGUPQNJJ-UHFFFAOYSA-N
Synonym 4,4-diamino-p-terphenyl, 4,4-diaminoterphenyl, 4-4-4-aminophenyl phenyl aniline, ccris 4965, p-terphenyl, 4,4-diamine, 1,1':4',1-terphenyl-4,4-diamine, 4'-4-aminophenyl-1,1'-biphenyl-4-amine, 4,4-diamino-4-terphenyl
PubChem CID 104949
IUPAC Name 4-[4-(4-aminophenyl)phenyl]aniline
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)N)C3=CC=C(C=C3)N

Specifications

Melting Point 244°C to 250°C
Quantity 1 g
Beilstein 2120295
Formula Weight 260.34
Percent Purity 95%
Chemical Name or Material 4,4''-Diamino-p-terphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'