missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminodiphenyl sulfone, 98%, Thermo Scientific Chemicals

Product Code. 11493208
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is active against a wide range of bacteria, but is mainly employed for its actions against Mycobacterium leprae, being used as part of multidrug regimens in the treatment of all forms of leprosy. It is a second-line medication for the treatment and prevention of Pneumocystis pneumonia and for the prevention of toxoplasmosis in those who have poor immune function. Additionally, it has been used for acne as well as other skin conditions. Moreover it is employed as an anti-inflammatory drug, anti-infective drug, a leprostatic drug and an antimalarial drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water. Solubility in hot methanol is almost transparent. Soluble in DMSO, ethanol and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-08-0
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD00007887
InChI Key MQJKPEGWNLWLTK-UHFFFAOYSA-N
Synonym dapsone, diaphenylsulfone, 4,4'-diaminodiphenyl sulfone, dapson, 4,4'-sulfonyldianiline, diaphenylsulfon, sulfona, 4-aminophenyl sulfone, avlosulfone, diphenasone
PubChem CID 2955
ChEBI CHEBI:4325
IUPAC Name 4-(4-aminophenyl)sulfonylaniline
SMILES C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 175°C to 178°C
Odor Odorless
Quantity 100 g
Beilstein 788055
Merck Index 14,2822
Solubility Information Insoluble in water. Solubility in hot methanol is almost transparent. Soluble in DMSO,ethanol and acetone.
Formula Weight 248.3
Percent Purity 98%
Chemical Name or Material 4,4'-Diaminodiphenyl sulfone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.