missing translation for 'onlineSavingsMsg'
Learn More

4,4″-Dibromo-p-terphenyl, 98%, Thermo Scientific Chemicals

Product Code. 15244418
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17788-94-2
Molecular Formula C18H12Br2
Molecular Weight (g/mol) 388.10
MDL Number MFCD00272889
InChI Key VAIPJQIPFPRJKJ-UHFFFAOYSA-N
Synonym 4,4-dibromo-p-terphenyl, 4,4-dibromo-1,1':4',1-terphenyl, 1,1':4',1-terphenyl, 4,4-dibromo, acmc-1c6h6, 1,4-bis 4-bromophenyl benzene, 4-bromo-4'-4-bromophenyl-1,1'-biphenyl, 1∼4∼,3∼4∼-dibromo-1∼1∼,2∼1∼:2∼4∼,3∼1∼-terphenyl, 4,4 inverted exclamation marka inverted exclamation marka-dibromo-p-terphenyl, 4,4 inverted exclamation marka inverted exclamation marka-dibromo-1,1 inverted exclamation marka:4 inverted exclamation marka,1 inverted exclamation marka inverted exclamation marka-terphenyl
PubChem CID 12455998
IUPAC Name 1,4-bis(4-bromophenyl)benzene
SMILES BrC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 318.0°C to 320.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 388.1
Percent Purity 98%
Chemical Name or Material 4,4''-Dibromo-p-terphenyl
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4''-Dibromo-p-terphenyl, 98%
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.