missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobiphenyl, 99%, Thermo Scientific Chemicals

31.60 € - 98.50 €

Chemical Identifiers

CAS 398-23-2
Molecular Formula C12H8F2
Molecular Weight (g/mol) 190.193
MDL Number MFCD00000349
InChI Key PZDAAZQDQJGXSW-UHFFFAOYSA-N
Synonym 4,4'-difluorobiphenyl, 4,4'-difluoro-1,1'-biphenyl, 4,4'-difluorodiphenyl, 1-fluoro-4-4-fluorophenyl benzene, unii-o0694gmk81, 1,1'-biphenyl, 4,4'-difluoro, 4-fluoro-1-4-fluorophenyl benzene, pubchem2016, 4,4'-difluorobipheny, biphenyl,4'-difluoro
PubChem CID 9811
IUPAC Name 1-fluoro-4-(4-fluorophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11308115
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L08361.03
1 g
31.60 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11318115
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L08361.06
5 g
98.50 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

398-23-2
190.193
PZDAAZQDQJGXSW-UHFFFAOYSA-N
9811
C1=CC(=CC=C1C2=CC=C(C=C2)F)F
C12H8F2
MFCD00000349
4,4'-difluorobiphenyl, 4,4'-difluoro-1,1'-biphenyl, 4,4'-difluorodiphenyl, 1-fluoro-4-4-fluorophenyl benzene, unii-o0694gmk81, 1,1'-biphenyl, 4,4'-difluoro, 4-fluoro-1-4-fluorophenyl benzene, pubchem2016, 4,4'-difluorobipheny, biphenyl,4'-difluoro
1-fluoro-4-(4-fluorophenyl)benzene

Specifications

398-23-2
254°C to 255°C
MFCD00000349
2044699
4,4'-difluorobiphenyl, 4,4'-difluoro-1,1'-biphenyl, 4,4'-difluorodiphenyl, 1-fluoro-4-4-fluorophenyl benzene, unii-o0694gmk81, 1,1'-biphenyl, 4,4'-difluoro, 4-fluoro-1-4-fluorophenyl benzene, pubchem2016, 4,4'-difluorobipheny, biphenyl,4'-difluoro
C1=CC(=CC=C1C2=CC=C(C=C2)F)F
190.193
190.19
4,4'-Difluorobiphenyl
87°C to 89°C
C12H8F2
UN3152
14,3149
PZDAAZQDQJGXSW-UHFFFAOYSA-N
1-fluoro-4-(4-fluorophenyl)benzene
9811
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only