missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxytrityl Chloride, 98%

Product Code. 10316550 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10316550

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40615-36-9 | C21H19ClO2 | 338.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40615-36-9
Molecular Formula C21H19ClO2
Molecular Weight (g/mol) 338.83
MDL Number MFCD00008409
InChI Key JBWYRBLDOOOJEU-UHFFFAOYSA-N
Synonym 4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl
PubChem CID 96831
IUPAC Name 1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 121°C to 127°C
Color Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H5C(C6H4OCH3)2Cl
Quantity 25 g
Fieser 01,188
Formula Weight 338.82
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Dimethoxytrityl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Dimethoxytrityl chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.