missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-1,3-cyclohexanedione, 98%, Thermo Scientific™
47.80 € - 161.90 €
Chemical Identifiers
CAS | 562-46-9 |
---|---|
Molecular Formula | C8H12O2 |
Molecular Weight (g/mol) | 140.18 |
MDL Number | MFCD00075224 |
InChI Key | PLGPBTCNKJQJHQ-UHFFFAOYSA-N |
PubChem CID | 136360 |
IUPAC Name | 4,4-dimethylcyclohexane-1,3-dione |
SMILES | CC1(C)CCC(=O)CC1=O |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
562-46-9 | |
140.18 | |
PLGPBTCNKJQJHQ-UHFFFAOYSA-N | |
4,4-dimethylcyclohexane-1,3-dione |
C8H12O2 | |
MFCD00075224 | |
136360 | |
CC1(C)CCC(=O)CC1=O |
Specifications
562-46-9 | |
100.0 | |
White | |
Authentic | |
C8H12O2 | |
4,4-dimethyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 4,4-dimethyl, 4,4-dimethyl-cyclohexane-1,3-dione, acmc-209lre, 4,4-dimethyl-1,3cyclohexanedione, 4,4dimethyl-1,3-cyclohexanedione, 4,4-dimethy 1,3-cyclohexanedione, 1,3-cyclohexanedione,4,4-dimethyl, 4,4-dimethyl-1,3-cyclohexanedione # | |
PLGPBTCNKJQJHQ-UHFFFAOYSA-N | |
4,4-dimethylcyclohexane-1,3-dione | |
136360 | |
98% | |
4, 4-Dimethyl-1, 3-cyclohexanedione |
97.5 | |
102°C to 105°C | |
130°C (7.0 mmHg) | |
97.5% min. (GC) | |
MFCD00075224 | |
Solubility in water: .. Other solubilities: soluble in acetone,halogenated solvents | |
CC1(C)CCC(=O)CC1=O | |
140.18 | |
140.18 | |
Needles |
Safety and Handling
Product Identifier
- 4,4-Dimethyl-1,3-cyclohexanedione