missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, 98%

Product Code. 10259383
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845-46-5
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.27
MDL Number MFCD00020350
InChI Key OSCKRHPYZNTEIO-UHFFFAOYSA-M
Synonym p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid
PubChem CID 23674498
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Quantity 2.5 g
Formula Weight 291.27
Percent Purity 98%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, 98%
Show More Show Less
Product Identifier
  • 4-(4-Dimethylaminophenylazo)benzoic acid sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.