missing translation for 'onlineSavingsMsg'
Learn More
4-(4-Ethylphenyl)-3-thiosemicarbazide, 98+%, Thermo Scientific™ Product Code.: 11338765

4-(4-Ethylphenyl)-3-thiosemicarbazide, 98+%, Thermo Scientific™

Product Code. 11338765
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11338765

Brand: Thermo Scientific Alfa Aesar L12361.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93693-01-7
Molecular Formula C9H13N3S
Molecular Weight (g/mol) 195.28
MDL Number MFCD00041303
InChI Key MXVVZBDSLDIVAW-UHFFFAOYSA-N
Synonym 4-4-ethylphenyl-3-thiosemicarbazide, n-4-ethylphenyl hydrazinecarbothioamide, 3-amino-1-4-ethylphenyl thiourea, 4-ethylphenyl amino hydrazinomethane-1-thione, 1-amino-3-4-ethylphenyl thiourea, 4-4'-ethylphenyl-3-thiosemicarbazideShow MoreShow Less
PubChem CID 3259670
IUPAC Name 1-amino-3-(4-ethylphenyl)thiourea
SMILES CCC1=CC=C(NC(=S)NN)C=C1

Specifica

Melting Point 128°C to 131°C
Quantity 1 g
UN Number UN2811
Beilstein 2804749
Formula Weight 195.29
Percent Purity ≥98%
Chemical Name or Material 4-(4-Ethylphenyl)-3-thiosemicarbazide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB1508 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
4-(4-Ethylphenyl)-3-thiosemicarbazide, 98+%, Thermo Scientific™ >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato