Learn More
4-(4-Fluorobenzoyl)pyridine, 99%, Thermo Scientific Chemicals
23.92 € - 353.76 €
Chemical Identifiers
CAS | 41538-36-7 |
---|---|
Molecular Formula | C12H8FNO |
Molecular Weight (g/mol) | 201.20 |
MDL Number | MFCD02683088 |
InChI Key | WTRWBYGUMQEFFI-UHFFFAOYSA-N |
Synonym | 4-4-fluorobenzoyl pyridine, 4-fluorophenyl pyridin-4-yl methanone, methanone, 4-fluorophenyl-4-pyridinyl, 4-fluorophenyl-pyridin-4-ylmethanone, 4-fluorophenyl 4-pyridyl ketone, acmc-20amw5, d0j0wo, 4-pyridyl p-fluorophenyl ketone, 4-4-fluorophenyl carbonyl pyridine, 4-fluorophenyl 4-pyridinyl methanone |
PubChem CID | 7023019 |
SMILES | FC1=CC=C(C=C1)C(=O)C1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11496026
|
Thermo Scientific Alfa Aesar
B22327.03 |
1 g |
23.92 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11406036
|
Thermo Scientific Alfa Aesar
B22327.06 |
5 g |
86.43 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416036
|
Thermo Scientific Alfa Aesar
B22327.14 |
25 g |
353.76 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-(4-Fluorobenzoyl)pyridine is used for increasing cardiac contractility and for the treatment of congestive heart failure, and pharmaceutical compositions and for the preparation thereof.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(4-Fluorobenzoyl)pyridine is used for increasing cardiac contractility and for the treatment of congestive heart failure, and pharmaceutical compositions and for the preparation thereof.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
41538-36-7 | |
201.20 | |
WTRWBYGUMQEFFI-UHFFFAOYSA-N | |
7023019 |
C12H8FNO | |
MFCD02683088 | |
4-4-fluorobenzoyl pyridine, 4-fluorophenyl pyridin-4-yl methanone, methanone, 4-fluorophenyl-4-pyridinyl, 4-fluorophenyl-pyridin-4-ylmethanone, 4-fluorophenyl 4-pyridyl ketone, acmc-20amw5, d0j0wo, 4-pyridyl p-fluorophenyl ketone, 4-4-fluorophenyl carbonyl pyridine, 4-fluorophenyl 4-pyridinyl methanone | |
FC1=CC=C(C=C1)C(=O)C1=CC=NC=C1 |
Specifications
41538-36-7 | |
C12H8FNO | |
4-4-fluorobenzoyl pyridine, 4-fluorophenyl pyridin-4-yl methanone, methanone, 4-fluorophenyl-4-pyridinyl, 4-fluorophenyl-pyridin-4-ylmethanone, 4-fluorophenyl 4-pyridyl ketone, acmc-20amw5, d0j0wo, 4-pyridyl p-fluorophenyl ketone, 4-4-fluorophenyl carbonyl pyridine, 4-fluorophenyl 4-pyridinyl methanone | |
FC1=CC=C(C=C1)C(=O)C1=CC=NC=C1 | |
201.20 | |
201.2 | |
4-(4-Fluorobenzoyl)pyridine |
85°C to 87°C | |
MFCD02683088 | |
WTRWBYGUMQEFFI-UHFFFAOYSA-N | |
4-(4-fluorobenzoyl)pyridine | |
7023019 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.