missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Iodophenoxy)tetrahydropyran, 97%, Thermo Scientific™

Product Code. 10614785
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 144808-72-0
Molecular Formula C11H13IO2
Molecular Weight (g/mol) 304.127
MDL Number MFCD09064966
InChI Key PQUFKWFIWPZFBM-UHFFFAOYSA-N
Synonym 4-4-iodophenoxy tetrahydropyran, 4-4-iodophenoxy oxane, 4-4-iodophenoxy tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-4-iodophenoxy, acmc-20e5io
PubChem CID 24229524
IUPAC Name 4-(4-iodophenoxy)oxane
SMILES C1COCCC1OC2=CC=C(C=C2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 304.12
Percent Purity 97%
Chemical Name or Material 4-(4-Iodophenoxy)tetrahydropyran
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.